Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Methoxyphenyl)piperazine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. M144925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M144925G 25 g
M14495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M144925G Supplier TCI America Supplier No. M144925G

Chemical Identifiers

CAS 38869-47-5
Molecular Formula C11H18Cl2N2O
Molecular Weight (g/mol) 265.178
MDL Number MFCD00012767
InChI Key MMXANKLJIHSIQH-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl piperazine dihydrochloride, 1-4-methoxyphenyl piperazinedihydrochloride, piperazine, 1-4-methoxyphenyl-, dihydrochloride, 1-4-methoxylphenyl-piperazine dihydrochloride, para-methoxyphenylpiperazine dihydrochloride, 1-4-methoxyphenyl-piperazine dihydrochloride, 4-methoxy-1-piperazinylbenzene, chloride, chloride, pubchem8579, acmc-1ct2c, ksc222g2n
PubChem CID 2723700
IUPAC Name 1-(4-methoxyphenyl)piperazine;dihydrochloride
SMILES COC1=CC=C(C=C1)N2CCNCC2.Cl.Cl

Specifications

Color Red-White
Quantity 25 g
UN Number 2811
Formula Weight 265.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Methoxyphenyl)piperazine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.