Learn More
1-(4-Methylphenyl)ethanol, 97%
Description
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.
Solubility
Slightly miscible with water.
Notes
Incompatible with oxidizing agents and strong reducing agents. Store in a cool place.
Specifications
Specifications
| Density | 0.987 |
| Boiling Point | 218°C to 220°C |
| Flash Point | 107°C (225°F) |
| Refractive Index | 1.523 |
| Quantity | 5 g |
| Beilstein | 1857562 |
| Merck Index | 14,3237 |
| Solubility Information | Slightly miscible with water. |
| Formula Weight | 136.2 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| EINECSNumber | 208-637-8 |
| RTECSNumber | DO4565000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.