Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Naphthalenebis(trifluoromethanesulfonate) 98.0+%, TCI America™
SDP

Catalog No. N09345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09345G 5 g
N09341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09345G Supplier TCI America Supplier No. N09345G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 152873-78-4
Molecular Formula C12H6F6O6S2
Molecular Weight (g/mol) 424.284
InChI Key WUQONKZHJZYVAX-UHFFFAOYSA-N
Synonym 1,4-Naphthaleneditriflate
PubChem CID 10526326
IUPAC Name [4-(trifluoromethylsulfonyloxy)naphthalen-1-yl] trifluoromethanesulfonate
SMILES C1=CC=C2C(=C1)C(=CC=C2OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 64°C
Color White
Quantity 5 g
Formula Weight 424.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Naphthalenebis(trifluoromethanesulfonate)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.