Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Naphthoquinone-2-sulfonic Acid Potassium Salt, Thermo Scientific™

Catalog No. AC415390050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC415390050 5 g
AC415390250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC415390050 Supplier Thermo Scientific Chemicals Supplier No. 415390050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34169-62-5
Molecular Formula C10H5KO5S
Molecular Weight (g/mol) 276.303
InChI Key WZEGLRXCWYMIDO-UHFFFAOYSA-M
Synonym potassium 1,4-naphthoquinone-2-sulfonate, 2-naphthalenesulfonic acid, 1,4-dihydro-1,4-dioxo-, potassium salt, potassium 1,4-dioxo-1,4-dihydronaphthalene-2-sulfonate, 1,4-naphthoquinone-2-sulfonic acid potassium salt, potassium 1,4-dioxonaphthalene-2-sulfonate, 2-naphthalenesulfonic acid, 1,4-dihydro-1,4-dioxo-, potassium salt 1:1, potassium 1,4-dihydro-1,4-dioxonaphthalene-2-sulphonate, 1,4-dioxo-2-naphthalenesulfonic acid potassium salt
PubChem CID 2735097
IUPAC Name potassium;1,4-dioxonaphthalene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C=C(C2=O)S(=O)(=O)[O-].[K+]

Specifications

Quantity 5 g
Formula Weight 276.3
Chemical Name or Material 1, 4-Naphthoquinone-2-sulfonic acid potassium salt
EINECSNumber 251-861-6
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.