Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Nitrophenyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. N069425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N069425G 25 g
N06945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N069425G Supplier TCI America Supplier No. N069425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6269-89-2
Molecular Formula C10H13N3O2
Molecular Weight (g/mol) 207.233
MDL Number MFCD00005961
InChI Key VWOJSRICSKDKAW-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl piperazine, 1-4-nitro-phenyl-piperazine, piperazine, 1-4-nitrophenyl, 1-4-nitrophenyl-piperazine, n-4-nitrophenyl piperazine, n-4-nitrophenyl-piperazine, 4-nitrophenyl piperazine, 1-4-nitrophenyl piperazin, pubchem8580, 4-nitrophenylpiperazine
PubChem CID 80447
SMILES C1CN(CCN1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 132°C
Color Yellow
Quantity 25 g
Formula Weight 207.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Nitrophenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.