Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Oxazepan-5-one, 97%

Click to view available options
Quantity:
1g
5g
25g
10341-26-1
C5H9NO2
115.132
MFCD02683091
OXDIYMHNQAWSCL-UHFFFAOYSA-N
tetrahydro-1,4-oxazepin-5 2h-one,1,4-oxazepin-5 2h-one, tetrahydro,4-oxacaprolactam,acmc-1bsly,tetrahydro-1,4-oxazepin-5 4h-one,tetrahydro-2h-1,4-oxazepin-5-one
7023020
1,4-oxazepan-5-one
C1COCCNC1=O
10341-26-1
C5H9NO2
115.132
MFCD02683091
OXDIYMHNQAWSCL-UHFFFAOYSA-N
tetrahydro-1,4-oxazepin-5 2h-one,1,4-oxazepin-5 2h-one, tetrahydro,4-oxacaprolactam,acmc-1bsly,tetrahydro-1,4-oxazepin-5 4h-one,tetrahydro-2h-1,4-oxazepin-5-one
7023020
1,4-oxazepan-5-one
C1COCCNC1=O
Applications
Used as a pharmaceutical intermediate.

Notes
Hygroscopic. Incompatible with oxidizing agents.
Quantity 1g
Melting Point 80°C to 82°C
Formula Weight 115.13
Percent Purity 97%
Chemical Name or Material 1,4-Oxazepan-5-one

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning