Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,4-Pentadiene, 99%
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | -148.0°C |
| Color | Colorless |
| Density | 0.6500g/mL |
| Boiling Point | 26.0°C |
| Flash Point | −20°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May be fatal if swallowed and enters airways. Extremely flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfac |
| EINECSNumber | 209-736-9 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction