Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Phenylenediamine Sulfate 98.0+%, TCI America™
SDP

Catalog No. P017525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P017525G 25 g
P0175500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P017525G Supplier TCI America Supplier No. P017525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16245-77-5
Molecular Formula C6H10N2O4S
Molecular Weight (g/mol) 206.22
MDL Number MFCD00035510
InChI Key UFPKLWVNKAMAPE-UHFFFAOYSA-N
Synonym p-phenylenediamine sulfate, benzene-1,4-diamine sulfate, 1,4-diaminobenzene sulfate, 1,4-phenylenediamine sulfate, 1,4-benzenediamine, sulfate 1:1, unii-s7igm30961, benzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, sulfate, benzene-1,4-diammonium sulphate, 1,4-benzenediamine sulfate
PubChem CID 27769
IUPAC Name benzene-1,4-diamine; sulfuric acid
SMILES OS(O)(=O)=O.NC1=CC=C(N)C=C1

Specifications

Color Purple-White
Quantity 25 g
UN Number 2811
Formula Weight 206.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Phenylenediamine Sulfate
EINECSNumber (3)-0185
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.