Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Piperazine Dialdehyde, MP Biomedicals

Catalog No. ICN227296 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN227296 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN227296 Supplier MP Biomedicals, Inc Supplier No. 0522729605

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4164-39-0
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00006153
InChI Key CBLGQEBXWDKYDI-UHFFFAOYSA-N
Synonym 1,4-diformylpiperazine, 1,4-piperazinedicarboxaldehyde, pubchem15032, acmc-1ago2, 1,4-piperazine dialdehyde, piperazine-1,4-dicarboxaldehyde, 1,4 piperazinedicarboxaldehyde
PubChem CID 77821
IUPAC Name piperazine-1,4-dicarbaldehyde
SMILES O=CN1CCN(CC1)C=O

Specifications

Quantity 5 g
Chemical Name or Material 1, 4-Piperazine Dialdehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.