Learn More
JDM SCIENTIFIC LLC 1.4ml external thread 2D Tube
Supplier:  JDM SCIENTIFIC LLC 204022
Encompass_Preferred
1 4ml TPX 2D barcoded external thread storage tube precapped with screw cap AlteClean
Catalog No. 50-000-14303
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.