Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Bis(methylamino)-3-oxapentane, 98%

Catalog No. AC275860010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275860010 1 g Glass bottle
AC275860050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC275860010 Supplier Thermo Scientific Chemicals Supplier No. 275860010
Add to Cart

CAS: 2620-27-1 | C6H16N2O | 132.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2620-27-1
Molecular Formula C6H16N2O
Molecular Weight (g/mol) 132.21
InChI Key VXPJBVRYAHYMNY-UHFFFAOYSA-N
PubChem CID 2724770
IUPAC Name N-methyl-2-[2-(methylamino)ethoxy]ethanamine
SMILES CNCCOCCNC

Specifications

Color Yellow
Density 0.8720g/mL
Boiling Point 76.0°C to 78.0°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4370 to 1.4390
Linear Formula CH3NHCH2CH2OCH2CH2NHCH3
Quantity 1 g
Specific Gravity 0.872
Synonym 2,2'-oxybis n-methylethanamine, ethanamine,2,2'-oxybis n-methyl, ethanamine, 2,2'-oxybis n-methyl, 1,5-bis methylamino-3-oxapentane, methyl 2-2-methylamino ethoxy ethyl amine, bmaop, n-methyl-2-methylaminoethyloxy ethylamine, n-methyl-2-2-methylamino ethoxy ethanamine, n-methyl-n-2-2-methylamino ethoxy ethyl amine
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,chloroform
Formula Weight 132.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 5-Bis(methylamino)-3-oxapentane
Show More Show Less
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.