Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1,5-Cyclooctadiene)(methoxy)iridium(I) Dimer, TCI America™
SDP

Catalog No. C26621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26621G 1 g
C2662200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26621G Supplier TCI America Supplier No. C26621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12148-71-9
Molecular Formula C18H30Ir2O2
Molecular Weight (g/mol) 662.87
MDL Number MFCD08459360
InChI Key SGBSCFWMCJNCRM-MIXQCLKLSA-N
Synonym bis 1,5-cyclooctadiene di-mu-methoxydiiridium i, 1,5-cyclooctadiene methoxy iridium i dimer
PubChem CID 124202702
IUPAC Name bis((1Z,5Z)-cycloocta-1,5-diene); bis(methoxyiridiumylium)
SMILES CO[Ir+].CO[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 179°C
Color Yellow
Quantity 1 g
Formula Weight 662.87
Physical Form Crystalline Powder
Chemical Name or Material (1,5-Cyclooctadiene)(methoxy)iridium(I) Dimer
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.