Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dibromo-2,6-dihydroxynaphthalene 98.0+%, TCI America™
SDP

Catalog No. D466825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D466825G 25 g
D46685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D466825G Supplier TCI America Supplier No. D466825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 132178-78-0
Molecular Formula C10H6Br2O2
Molecular Weight (g/mol) 317.96
MDL Number MFCD16251539
InChI Key COJNHIANORGBGY-UHFFFAOYSA-N
Synonym 1,5-Dibromo-2,6-naphthalenediol
PubChem CID 14896989
IUPAC Name 1,5-dibromonaphthalene-2,6-diol
SMILES OC1=CC=C2C(Br)=C(O)C=CC2=C1Br

Specifications

Melting Point 215°C
Quantity 25 g
Formula Weight 317.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dibromo-2,6-dihydroxynaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.