Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Diisocyanatonaphthalene 98.0+%, TCI America™
SDP

Catalog No. N0168100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0168100G 100 g
N016825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0168100G Supplier TCI America Supplier No. N0168100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3173-72-6
Molecular Formula C12H6N2O2
Molecular Weight (g/mol) 210.192
InChI Key SBJCUZQNHOLYMD-UHFFFAOYSA-N
Synonym Naphthalene 1,5-Diisocyanate
PubChem CID 18503
ChEBI CHEBI:82496
IUPAC Name 1,5-diisocyanatonaphthalene
SMILES C1=CC2=C(C=CC=C2N=C=O)C(=C1)N=C=O

Specifications

Melting Point 129°C
Color White-Yellow
Boiling Point 183°C
Quantity 100 g
UN Number 2206
Formula Weight 210.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diisocyanatonaphthalene
EINECSNumber (4)-0527
RTECSNumber NQ9600000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.