Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dimercaptonaphthalene 98.0+%, TCI America™
SDP

Catalog No. D21821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21821G 1 g
D21825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21821G Supplier TCI America Supplier No. D21821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5325-88-2
Molecular Formula C10H8S2
Molecular Weight (g/mol) 192.294
MDL Number MFCD00068909
InChI Key AAPAGLBSROJFGM-UHFFFAOYSA-N
Synonym 1,5-Naphthalenedithiol
PubChem CID 21397
IUPAC Name naphthalene-1,5-dithiol
SMILES C1=CC2=C(C=CC=C2S)C(=C1)S

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 1 g
Formula Weight 192.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dimercaptonaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.