Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dinitronaphthalene 98.0+%, TCI America™
SDP

Catalog No. D0836250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0836250G 250 g
D083625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0836250G Supplier TCI America Supplier No. D0836250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 605-71-0
Molecular Formula C10H6N2O4
Molecular Weight (g/mol) 218.168
MDL Number MFCD00003916
InChI Key ZUTCJXFCHHDFJS-UHFFFAOYSA-N
Synonym naphthalene, 1,5-dinitro, naphthalene, dinitro, unii-ui2s14iw4t, ccris 6028, ui2s14iw4t, napththalene, 1,5-dinitro, 1,5 or 1,8-dinitronaphthalene, acmc-1bixi, 1,5-dinitro naphthalene, 1,5-dinitro-naphthalene
PubChem CID 11803
ChEBI CHEBI:50640
IUPAC Name 1,5-dinitronaphthalene
SMILES C1=CC2=C(C=CC=C2[N+](=O)[O-])C(=C1)[N+](=O)[O-]

Specifications

Melting Point 218°C
Color Yellow
Quantity 250 g
Formula Weight 218.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dinitronaphthalene
EINECSNumber (4)-0325
RTECSNumber QJ4551000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.