Learn More
1,5-Diphenylcarbazide, ACS
Description
1,5-Diphenylcarbazide is used in the colorimetric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 170°C to 176°C |
| Color | White to Cream |
| Quantity | 5 g |
| Beilstein | 752039 |
| Sensitivity | Air and light sensitive |
| Merck Index | 14,3321 |
| Solubility Information | Soluble in glacial acetic acid,alcohol and acetone; Slightly soluble in water. |
| Formula Weight | 242.28 |
| Grade | ACS |
| Physical Form | Crystalline |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 205-403-7 |
| RTECSNumber | FF2750000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.