Learn More
Sigma Organic Chemistry 1- 5-Methoxy-1 3-benzox | 1G | MFCD11696398
Supplier: Sigma Organic Chemistry CBR010621G

1- 5-Methoxy-1 3-benzox , 1G
About this Item:
MDL #: MFCD11696398
Molecular Weight: 214.65
UNSPSC Code: 12352200
Chemical Formula: C9H11ClN2O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.