Learn More
Sigma Organic Chemistry 1- 5-Methyl-1H-benzimid | 1G | MFCD11656750
Supplier: Sigma Organic Chemistry CBR012761G

1- 5-Methyl-1H-benzimid , 1G
About this Item:
MDL #: MFCD11656750
Molecular Weight: 248.15
UNSPSC Code: 12352200
Chemical Formula: C10H15Cl2N3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.