Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Naphthalenebis(trifluoromethanesulfonate) 98.0+%, TCI America™
SDP

Catalog No. N09255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N09255G Supplier TCI America Supplier No. N09255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 152873-79-5
Molecular Formula C12H6F6O6S2
Molecular Weight (g/mol) 424.28
MDL Number MFCD23380211
InChI Key IFHGSECJTKJJIJ-UHFFFAOYSA-N
Synonym 1,5-Naphthaleneditriflate
PubChem CID 14976571
IUPAC Name 5-(trifluoromethanesulfonyloxy)naphthalen-1-yl trifluoromethanesulfonate
SMILES FC(F)(F)S(=O)(=O)OC1=CC=CC2=C(OS(=O)(=O)C(F)(F)F)C=CC=C12

Specifications

Melting Point 115°C
Color White
Quantity 5 g
Formula Weight 424.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Naphthalenebis(trifluoromethanesulfonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.