Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(5-(Trifluoromethyl)-2-pyridyl)piperidine-4-carboxylic acid, 95%, Thermo Scientific™

Catalog No. KM07694CB
Change view
Click to view available options
Quantity:
250 mg
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM07694CB 250 mg
KM07694EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. KM07694CB Supplier Thermo Scientific Chemicals Supplier No. KM07694CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 406476-31-1
Molecular Formula C12H13F3N2O2
Molecular Weight (g/mol) 274.24
MDL Number MFCD00209248
InChI Key KNDSIDUPVUCATQ-UHFFFAOYSA-N
Synonym 1-5-trifluoromethyl pyridin-2-yl piperidine-4-carboxylic acid, 1-5-trifluoromethyl-2-pyridyl piperidine-4-carboxylic acid, 5'-trifluoromethyl-3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-4-carboxylic acid, peakdale1_002677, 2-4-carboxypiperidin-1-yl-5-trifluoromethyl pyridine, 4-carboxy-1-5-trifluoromethyl pyridin-2-yl piperidine, 1-5-trifluoromethyl pyridin-2-yl piperidine-4-carboxylicacid, 1-5-trifluoromethylpyrid-2-yl-piperidine-4-carboxylic acid
PubChem CID 2777788
SMILES [O-]C(=O)C1CCN(CC1)C1=CC=C(C=[NH+]1)C(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name 2-(4-carboxylatopiperidin-1-yl)-5-(trifluoromethyl)pyridin-1-ium
Formula Weight 274.24
Percent Purity 95%
Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridyl]piperidine-4-carboxylic acid
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.