Learn More
Thomas Scientific 1.5 mL,TUBE,NATURAL
SDP
Supplier:  Thomas Scientific 1149X75
Encompass_Preferred
1.5ml Natural Low Retention Microtubes, Phenix Brand, 250/Unit, PP, Certified RNase/DNase and DNA Free. This tube is not boil-proof.
Catalog No. NC0844503