Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Dimethoxyhexane 97.0+%, TCI America™
SDP

Catalog No. D28585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28585G Supplier TCI America Supplier No. D28585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13179-98-1
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00142569
InChI Key GZHQUHIMIVEQFV-UHFFFAOYSA-N
Synonym hexane,1,6-dimethoxy, hexamethylene glycol dimethyl ether, hexane, 1,6-dimethoxy, acmc-1c0t4, 1,6-dimethoxyhexane
PubChem CID 542359
IUPAC Name 1,6-dimethoxyhexane
SMILES COCCCCCCOC

Specifications

Color Colorless
Quantity 5 g
Formula Weight 146.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Dimethoxyhexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.