Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,7-Diaminoheptane 98.0+%, TCI America™
SDP

Catalog No. D009425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D009425G 25 g
D00945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D009425G Supplier TCI America Supplier No. D009425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 646-19-5
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.24
MDL Number MFCD00008246
InChI Key PWSKHLMYTZNYKO-UHFFFAOYSA-N
Synonym 1,7-diaminoheptane, 1,7-heptanediamine, heptamethylenediamine, chembl28242, h2n ch2 7nh2, heptan-1,7-diamin, 1,9-diazanonane, acmc-1atsg, 1,7-heptamethylenediamine
PubChem CID 69533
IUPAC Name heptane-1,7-diamine
SMILES NCCCCCCCN

Specifications

Color White
Boiling Point 225°C
Quantity 25 g
UN Number 3259
Formula Weight 130.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,7-Diaminoheptane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.