Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,7-Dichloro-1,1,3,3,5,5,7,7-octamethyltetrasiloxane 97.0+%, TCI America™
SDP

Catalog No. D526725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D526725G 25 g
D52675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D526725G Supplier TCI America Supplier No. D526725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2474-02-4
Molecular Formula C8H24Cl2O3Si4
Molecular Weight (g/mol) 351.52
MDL Number MFCD00045148
InChI Key UHRAUGIQJXURFE-UHFFFAOYSA-N
PubChem CID 75584
IUPAC Name chloro-[[[chloro(dimethyl)silyl]oxy-dimethylsilyl]oxy-dimethylsilyl]oxy-dimethylsilane
SMILES C[Si](C)(O[Si](C)(C)O[Si](C)(C)Cl)O[Si](C)(C)Cl

Specifications

Melting Point -62°C
Color Colorless
Boiling Point 222°C
Quantity 25 g
UN Number 2987
Formula Weight 351.52
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,7-Dichloro-1,1,3,3,5,5,7,7-octamethyltetrasiloxane
EINECSNumber (7)-0472
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.