Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,7-Heptanedioic acid, 98%, Thermo Scientific™

Catalog No. AA3952309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3952309 10 g
AA3952318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3952309 Supplier Thermo Scientific Chemicals Supplier No. 3952309

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-16-0
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004425
InChI Key WLJVNTCWHIRURA-UHFFFAOYSA-N
Synonym pimelic acid, 1,5-pentanedicarboxylic acid, pileric acid, heptandioic acid, heptane-1,7-dioic acid, pimelate, 6-carboxyhexanoate, 6-carboxyhexanoic acid, 1,7-heptanedioic acid, unii-bzq96wx25f
PubChem CID 385
ChEBI CHEBI:30531
IUPAC Name heptanedioic acid
SMILES OC(=O)CCCCCC(O)=O

Specifications

Melting Point 103°C to 105°C
Odor Odorless
Quantity 10 g
Merck Index 14,7431
Solubility Information Soluble in alcohol, ether
Formula Weight 160.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,7-Heptanedioic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.