Learn More
Sigma Organic Chemistry 1 7-Heptanediol | 25G | 629-30-1 | MFCD00002987
Supplier: Sigma Organic Chemistry H220125G

1,7-Heptanediol, 25G
About This Item:
Synonym(s): Heptamethylene glycol
Linear Formula: HO(CH2)7OH
Molecular Weight: 132.2
CAS Number: 629-30-1
MDL Number: MFCD00002987
UNSPSC Code: 12352100
Purity: 0.95
Boiling Point: 259 C (lit.)
Melting Point: 17-19 C (lit.)
Density: 0.951 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.455 (lit.)
Storage: room temp
EINECS Number: 211-085-0
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.