Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Bis(dimethylamino)naphthalene 98.0+%, TCI America™
SDP

Catalog No. B101825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B101825G 25 g
B10181G 1 g
B10185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B101825G Supplier TCI America Supplier No. B101825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20734-58-1
Molecular Formula C14H18N2
Molecular Weight (g/mol) 214.312
MDL Number MFCD00003920
InChI Key GJFNRSDCSTVPCJ-UHFFFAOYSA-N
Synonym 1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine
PubChem CID 88675
IUPAC Name 1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine
SMILES CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C

Specifications

Melting Point 48°C
Boiling Point 145°C
Quantity 25 g
Formula Weight 214.31
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Bis(dimethylamino)naphthalene [for Dehydrohalogenation]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.