Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diaminonaphthalene 99%, Thermo Scientific™

Catalog No. AC112301000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC112301000 100 g
AC112305000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112301000 Supplier Thermo Scientific Chemicals Supplier No. 112301000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-27-6
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
InChI Key YFOOEYJGMMJJLS-UHFFFAOYSA-N
Synonym 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t
PubChem CID 68067
IUPAC Name naphthalene-1,8-diamine
SMILES C1=CC2=C(C(=C1)N)C(=CC=C2)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 63°C to 66°C
Color Brown to Gray-Red
Infrared Spectrum Authentic
Linear Formula C10H6(NH2)2
Quantity 100 g
Beilstein 13, 205
Merck Index 15, 6457
Formula Weight 158.2
Percent Purity ≥98.5%
Physical Form Crystalline Solid
Chemical Name or Material 1, 8-Diaminonaphthalene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 207-529-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.