Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diaminonaphthalene 98.0+%, TCI America™
SDP

Catalog No. D0102100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0102100G 100 g
D010225G 25 g
D0102500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0102100G Supplier TCI America Supplier No. D0102100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 479-27-6
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00004033
InChI Key YFOOEYJGMMJJLS-UHFFFAOYSA-N
Synonym 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t
PubChem CID 68067
IUPAC Name naphthalene-1,8-diamine
SMILES C1=CC2=C(C(=C1)N)C(=CC=C2)N

Specifications

Melting Point 65°C
Boiling Point 205°C
Quantity 100 g
Formula Weight 158.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Diaminonaphthalene
EINECSNumber (4)-0324
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.