Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diazabicyclo[5.4.0]-7-undecene 98.0+%, TCI America™
SDP

Catalog No. D1270100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1270100G 100 g
D127025G 25 g
D1270500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1270100G Supplier TCI America Supplier No. D1270100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6674-22-2
Molecular Formula C9H16N2
Molecular Weight (g/mol) 152.241
MDL Number MFCD00006930
InChI Key GQHTUMJGOHRCHB-UHFFFAOYSA-N
Synonym 1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
PubChem CID 81184
IUPAC Name 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
SMILES C1CCC2=NCCCN2CC1

Specifications

Melting Point -70°C
Color Yellow
pH 12.8 (10g/L H2O soln. 20°C)
Boiling Point 83°C
Quantity 100 g
UN Number 3267
Formula Weight 152.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,8-Diazabicyclo[5.4.0]-7-undecene
EINECSNumber (5)-1117
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.