Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Dibromohexadecafluorooctane 97.0+%, TCI America™
SDP

Catalog No. D35871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35871G 1 g
D35875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35871G Supplier TCI America Supplier No. D35871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 812-58-8
Molecular Formula C8Br2F16
Molecular Weight (g/mol) 559.87
MDL Number MFCD00153111
InChI Key LYRJPHOHVRHNQH-UHFFFAOYSA-N
Synonym 1,8-Dibromoperfluorooctane
PubChem CID 2736786
IUPAC Name 1,8-dibromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctane
SMILES FC(F)(Br)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Specifications

Melting Point 39°C
Color White
Boiling Point 180°C
Quantity 1 g
Formula Weight 559.87
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,8-Dibromohexadecafluorooctane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.