Learn More
Sigma Organic Chemistry 1 8-Dibromooctane | 25G | 4549-32-0 | MFCD00000277
Supplier: Sigma Organic Chemistry D4260725G

1,8-Dibromooctane, 25G
About This Item:
Synonym(s): Octamethylene dibromide
Linear Formula: Br(CH2)8Br
Molecular Weight: 272.02
CAS Number: 4549-32-0
MDL Number: MFCD00000277
UNSPSC Code: 12352100
Purity: 0.98
Boiling Point: 270-272 C (lit.)
Melting Point: 12-16 C (lit.)
Density: 1.477 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.498 (lit.)
Storage: room temp
EINECS Number: 224-912-5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.