Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Dichloroanthraquinone 95.0+%, TCI America™
SDP

Catalog No. D0329250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0329250G 250 g
D032925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0329250G Supplier TCI America Supplier No. D0329250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82-43-9
Molecular Formula C14H6Cl2O2
Molecular Weight (g/mol) 277.1
MDL Number MFCD00001191
InChI Key VBQNYYXVDQUKIU-UHFFFAOYSA-N
Synonym 1,8-dichloroanthraquinone, 9,10-anthracenedione, 1,8-dichloro, 1,8-dichloranthrachinon, 1,8-dichloro-9,10-anthraquinone, anthraquinone, 1,8-dichloro, 1,8-dichloranthrachinon czech, acmc-1bmbx, 9, 1,8-dichloro, anthraquinone,8-dichloro
PubChem CID 6708
IUPAC Name 1,8-dichloroanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)Cl)C(=O)C3=C(C2=O)C=CC=C3Cl

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 250 g
Formula Weight 277.10
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Dichloroanthraquinone
EINECSNumber (4)-0689
RTECSNumber CB6496000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.