Learn More
1,8-Naphthalic anhydride, 97%
Description
1,8-Naphthalic anhydride is used as a seed protectant and to prepare dyestuffs and optical brighteners. It is used in the preparation of napthalimide analogues, which finds application as anticancer agents. Further, it is used as fluorescent brightening agent for polymeric materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,8-Naphthalic anhydride is used as a seed protectant and to prepare dyestuffs and optical brighteners. It is used in the preparation of napthalimide analogues, which finds application as anticancer agents. Further, it is used as fluorescent brightening agent for polymeric materials.
Solubility
Soluble in ether and alcohol. Slightly soluble in acetic acid. Insoluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Specifications
| Melting Point | 269°C to 273°C |
| Quantity | 1000 g |
| Beilstein | 153190 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in ether and alcohol. Slightly soluble in acetic acid. Insoluble in water. |
| Formula Weight | 198.18 |
| Percent Purity | 97% |
| Chemical Name or Material | 1,8-Naphthalic anhydride |
Safety and Handling
| Hazard Category | H317-H319 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c |
| EINECSNumber | 201-380-2 |
| RTECSNumber | QK5350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.