Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Naphthalimide, 98%

Catalog No. AAA1885630
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1885630 250 g
AAA1885609 10 g
AAA1885618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1885630 Supplier Thermo Scientific Chemicals Supplier No. A1885630

CAS: 81-83-4 | C12H7NO2 | 197.193 g/mol

It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

Solubility
Insoluble in water.

Notes
Store away from and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-83-4
Molecular Formula C12H7NO2
Molecular Weight (g/mol) 197.193
MDL Number MFCD00006920
InChI Key XJHABGPPCLHLLV-UHFFFAOYSA-N
Synonym 1,8-naphthalimide, naphthalimide, 1h-benz de isoquinoline-1,3 2h-dione, 1h-benzo de isoquinoline-1,3 2h-dione, 1,8-naphthalenedicarboximide, benzo de isoquinoline-1,3-dione, unii-gi0tv19gbn, naphthalene-1,8-dicarboximide, gi0tv19gbn
PubChem CID 66491
IUPAC Name benzo[de]isoquinoline-1,3-dione
SMILES C1=CC2=C3C(=C1)C(=O)NC(=O)C3=CC=C2

Specifications

Melting Point 299°C to 303°C
Quantity 250 g
Beilstein 153150
Solubility Information Insoluble in water.
Formula Weight 197.2
Percent Purity 98%
Chemical Name or Material 1,8-Naphthalimide
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 201-379-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.