Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Nonadiene 98.0+%, TCI America™
SDP

Catalog No. N033225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N033225ML 25 mL
N03325ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N033225ML Supplier TCI America Supplier No. N033225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4900-30-5
Molecular Formula C9H16
Molecular Weight (g/mol) 124.227
MDL Number MFCD00008669
InChI Key VJHGSLHHMIELQD-UHFFFAOYSA-N
Synonym 1,8-nonadiene, acmc-209kep
PubChem CID 78612
IUPAC Name nona-1,8-diene
SMILES C=CCCCCCC=C

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 mL
UN Number 3295
Formula Weight 124.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,8-Nonadiene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.