Learn More
Accuflow Systems Saint-Gobain SaniPure BDF Tubing, TPE, 1/8 in ID, 50 ft
Supplier:  Accuflow Systems AR400007
Encompass_Preferred
SaniPure BDF biopharmaceutical tubing is an animal-origin-free thermoplastic elastomer (TPE) designed for sterile fluid transfer and processing in pharmaceutical and laboratory environments. The tubing provides an ultra-smooth inner bore, low extractables and adsorption, and compatibility with standard sterilization and sterile-welding processes, making it well suited for media transfer, filling, and bioreactor lines.
  • Animal-origin-free thermoplastic elastomer material.
  • 1/8 in inner diameter, 1/4 in outer diameter, 50 ft nominal length.
  • Ultra-smooth inner bore to reduce particle entrapment.
  • Low extractables and adsorption compared with silicone tubing.
  • Meets USP Class VI and European Pharmacopoeia biocompatibility criteria.
  • Autoclavable and compatible with common sterilization and sterile-welding methods.
  • Good peristaltic pump life and heat-sealable for custom assemblies.
Catalog No. NC2887427