Learn More
Cole-Parmer Thermo Scientific Nunc 377267 CryoTubes, Internal Thread/Starfoot/1.8 mL; 50/Pk
Supplier:  Cole-Parmer 0375762
Encompass_Preferred
Presterilized by cobalt-60 radiation, these one-piece molded polypropylene vials withstand temperatures from -196 to 135°C. Starfooted vials lock into CryoTube vial holder 03755-71 for one-handed operation. Optional colored cap inserts simplify sample identification. Note: Do not autoclave or immerse vials in liquid nitrogen. For storage under extreme conditions such as liquid nitrogen, use CryoFlex™ tubing 03755-64.
Catalog No. NC0585083
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.