Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,9-Decadiene 98.0+%, TCI America™
SDP

Catalog No. D024525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D024525ML 25 mL
D02455ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D024525ML Supplier TCI America Supplier No. D024525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1647-16-1
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00008670
InChI Key NLDGJRWPPOSWLC-UHFFFAOYSA-N
Synonym 1,9-decadiene, decadiene-1,9, unii-2kwz01g244, acmc-1c2oy, dsstox_cid_2159, dsstox_rid_76506, dsstox_gsid_22159, chembl31020
PubChem CID 15439
IUPAC Name deca-1,9-diene
SMILES C=CCCCCCCC=C

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 mL
UN Number 3295
Formula Weight 138.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,9-Decadiene
EINECSNumber (2)-3625
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.