Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetoxy-3-chloroacetone 95.0+%, TCI America™
SDP

Catalog No. A123225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A123225G 25 g
A12325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A123225G Supplier TCI America Supplier No. A123225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40235-68-5
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00191365
InChI Key LCMPIWIWBGHBOY-UHFFFAOYSA-N
Synonym 1-Acetoxy-3-acetenyl Chloride, 1-Acetoxy-3-chloro-2-propanone
PubChem CID 38413
IUPAC Name 3-chloro-2-oxopropyl acetate
SMILES CC(=O)OCC(=O)CCl

Specifications

Color Yellow
Quantity 25 g
UN Number 1760
Formula Weight 150.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Acetoxy-3-chloroacetone
RTECSNumber AF8600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.