Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetyl-2-pyrrolidone 98.0+%, TCI America™
SDP

Catalog No. A141825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A141825G 25 g
A14185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A141825G Supplier TCI America Supplier No. A141825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 932-17-2
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.143
MDL Number MFCD00014101
InChI Key YLHUPYSUKYAIBW-UHFFFAOYSA-N
PubChem CID 70266
IUPAC Name 1-acetylpyrrolidin-2-one
SMILES CC(=O)N1CCCC1=O

Specifications

Boiling Point 231°C
Quantity 25 g
Formula Weight 127.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Acetyl-2-pyrrolidone
RTECSNumber UY5717000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.