Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetyl-4-(4-hydroxyphenyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. A1845250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1845250G 250 g
A184525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1845250G Supplier TCI America Supplier No. A1845250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67914-60-7
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.27
MDL Number MFCD00044905
InChI Key AGVNLFCRZULMKK-UHFFFAOYSA-N
Synonym 1-acetyl-4-4-hydroxyphenyl piperazine, 4-1-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, n-acetyl-4-4-hydroxyphenyl piperazine, 4-4-acetyl-1-piperazinyl phenol, piperazine, 1-acetyl-4-4-hydroxyphenyl, 4-4-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethan-1-one, 1-acetyl-4-4-hydroxylphenyl piperazine
PubChem CID 712441
IUPAC Name 1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethan-1-one
SMILES CC(=O)N1CCN(CC1)C1=CC=C(O)C=C1

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 250 g
Formula Weight 220.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.