Learn More
1-Acetylnaphthalene, 97+%
Description
They are used as ingredients of fragrances. Their derivatives are used as anti-tubercular agents. The end applications include soap, detergent, beauty care product, household product. It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
They are used as ingredients of fragrances. Their derivatives are used as anti-tubercular agents. The end applications include soap, detergent, beauty care product, household product. It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
Solubility
Not miscible or difficult to mix in water. Soluble in ethyl alcohol. Insoluble in paraffin oil.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 9°C to 11°C |
| Density | 1.12 |
| Boiling Point | 302°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.628 |
| Quantity | 1000 g |
| Beilstein | 1100618 |
| Solubility Information | Not miscible or difficult to mix in water. Soluble in ethyl alcohol. Insoluble in paraffin oil. |
| Formula Weight | 170.21 |
| Percent Purity | ≥97% |
| Show More |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 213-384-1 |
| RTECSNumber | AL2982800 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.