Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetylpiperazine 98.0+%, TCI America™
SDP

Catalog No. A139925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A139925G 25 g
A13995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A139925G Supplier TCI America Supplier No. A139925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13889-98-0
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.175
MDL Number MFCD00058676
InChI Key PKDPUENCROCRCH-UHFFFAOYSA-N
Synonym 1-acetylpiperazine, n-acetylpiperazine, 1-piperazin-1-yl ethanone, 1-piperazin-1-yl ethan-1-one, n-acetyl piperazine, acetylpiperazine, 1-acetyl piperazine, piperazine, 1-acetyl, 1-piperazin-1-yl-ethanone, 1-piperazin-1-yl ethanon
PubChem CID 83795
IUPAC Name 1-piperazin-1-ylethanone
SMILES CC(=O)N1CCNCC1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 128.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Acetylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.