Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-1-Acid Glycoprotein from Human Plasma, MP Biomedicals™

Catalog No. MP219134801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219134801 1 mg
MP219134805 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219134801 Supplier MP Biomedicals, Inc Supplier No. 0219134801

α-1-Acid Glycoprotein from Human Plasma ≥95%, White Powder

  • It is used for the study of the structure of oligosaccharide units.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66455-27-4
MDL Number MFCD00131224
Synonym a1-AG, orosomucoid, a1-seromucoid, AGP

Specifications

Color White
Quantity 1 mg
Physical Form Powder
Grade >95%
storageNote1 -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.