Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanamine hydrochloride, 99%

Catalog No. AAA1269922
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1269922 100 g
AAA1269914 25 g
AAA1269936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1269922 Supplier Thermo Scientific Chemicals Supplier No. A1269922
Add to Cart

CAS: 665-66-7 | C10H18ClN | 187.71 g/mol

1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.

Solubility
Soluble in water (50 mg/ml), ethanol (38 mg/ml at 25°C), chloroform (1:18), DMSO (38 mg/ml at 25°C), and methanol

Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 665-66-7
Molecular Formula C10H18ClN
Molecular Weight (g/mol) 187.71
MDL Number MFCD00074723
InChI Key WOLHOYHSEKDWQH-UHFFFAOYSA-N
Synonym amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane
PubChem CID 64150
ChEBI CHEBI:2619
IUPAC Name adamantan-1-amine;hydrochloride
SMILES [H+].[Cl-].NC12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 360°C
Quantity 100 g
Beilstein 4198854
Merck Index 14374
Solubility Information Soluble in water (50mg/ml),ethanol (38mg/ml at 25°C),chloroform (1:18),DMSO (38mg/ml at 25°C),and methanol
Formula Weight 187.71
Percent Purity 99%
Chemical Name or Material 1-Adamantanamine hydrochloride
Hazard Category Acute toxicity (category 4)
Hazard Statement Harmful if swallowed
EINECSNumber 211-560-2
RTECSNumber AU4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.