Learn More
Sigma Organic Chemistry 1-Allyl-1H-benzimidazol | 10mg | MFCD04355428
Supplier: Sigma Organic Chemistry CDS00696310MG

1-Allyl-1H-benzimidazol, 10mg
About this Item:
MDL #: MFCD04355428
Molecular Weight: 186.21
UNSPSC Code: 12352200
Chemical Formula: C11H10N2O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.