Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Allyl-3-methylimidazolium Chloride 98.0+%, TCI America™
SDP

Catalog No. A208425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A208425G 25 g
A20845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A208425G Supplier TCI America Supplier No. A208425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65039-10-3
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.629
MDL Number MFCD07784450
InChI Key QVRCRKLLQYOIKY-UHFFFAOYSA-M
Synonym 1-allyl-3-methylimidazolium chloride, 1-allyl-3-methyl-1h-imidazol-3-ium chloride, 1-methyl-3-prop-2-en-1-yl imidazol-1-ium chloride, amim-cl, 1-allyl-3-methylimidazoliumchloride, dsstox_cid_27904, dsstox_rid_82657, dsstox_gsid_47928, 1-methyl-3-allylimidazolium chloride, 1-allyl-3-methyl-3-imidazolium chloride
PubChem CID 11321106
IUPAC Name 1-methyl-3-prop-2-enylimidazol-1-ium;chloride
SMILES C[N+]1=CN(C=C1)CC=C.[Cl-]

Specifications

Quantity 25 g
Formula Weight 158.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Allyl-3-methylimidazolium Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.