Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-2-Naphthol-4-Sulfonic Acid Solution, For Phosphorus, Certified, LabChem™

Catalog No. LC108607 Shop All LabChem Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC108607 125 mL
LC108601 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC108607 Supplier LabChem, Inc. Supplier No. LC108607
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7681-57-4 , 7757-83-7 , 116-63-2
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Color Yellow
CAS Max % 90, 6.2, 3.7, 0.1
Linear Formula H2N(HO)C10H5SO3H
Solubility Information Soluble in water
Formula Weight 239.26
Grade Certified
Identification Passes Test
Packaging White Poly
Suitability Passes Test
Decomposition Information Sulfur compounds
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material 1-Amino-2-Naphthol-4-Sulfonic Acid Solution, For Phosphorus
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Take off contaminated clothing and wash before reuse.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.